6-Bromo-2-methoxyquinoline 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H54226-03
H54226-06
AAH54226-03EA
160
USD
AAH54226-03
AAH54226-06
AAH54226-MD
6-Bromo-2-methoxyquinoline 96%
6-Bromo-2-methoxyquinoline
Purity: 96%
CAS No.: 99455-05-7
Molecular Formula: C10H8BrNO
Formula Weight: 238.08
Melting Point: 87-89°
MDL No.: MFCD11847805
Formula:
C₁₀H₈BrNO MW: 238.08 g/mol Melting Pt: 87…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD11847805 CAS Number: 99455-05-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...