2-(5-Norbornene-2,3-dicarboximido)-1,1,3,3-tetramethyluronium Tetrafluoroborate ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: TNTU, 2-(1,3-Dioxo-3a,4,7,7a-tetrahydro-1H-4,7-methanoisoindol-2(3H)-yl)-1,1,3,3-tetramethyluronium tetrafluoroborate

H54194-03 H54194-06 H54194-14
AAH54194-03EA 39.6 USD
AAH54194-03 AAH54194-06 AAH54194-14
2-(5-Norbornene-2,3-dicarboximido)-1,1,3,3-tetramethyluronium Tetrafluoroborate ≥98%
2-(5-Norbornene-2,3-dicarboximido)-1,1,3,3-tetramethyluronium tetrafluoroborate

Purity: 98+% CAS No.: 125700-73-4 Molecular Formula: C14H20BF4N3O3 Formula Weight: 365.13 Storage Temperature: 2°C to 8°C Melting Point: 216-219° MDL No.: MFCD00167753

Formula: C₁₄H₂₀BF₄N₃O₃
Melting Pt: 216…219 °C
Storage Temperature: Ambient
MDL Number: MFCD00167753
CAS Number: 125700-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR