5-Fluoroindoline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H54158-03
H54158-06
H54158-MD
AAH54158-03EA
79
USD
AAH54158-03
AAH54158-06
AAH54158-MD
5-Fluoroindoline 97%
5-Fluoroindoline
Purity: 97%
CAS No.: 2343-22-8
Molecular Formula: C8H8FN
Formula Weight: 137.16
Storage Temperature: 2°C to 8°C
Formula:
C₈H₈FN MW: 137.16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00214461 CAS Number: 2343-22-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...