2-(Ethylcarbamoyl)pyridine-5-boronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Ethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinamide

H54151-03 H54151-MD
AAH54151-03EA 162 USD
AAH54151-03 AAH54151-MD
2-(Ethylcarbamoyl)pyridine-5-boronic acid pinacol ester 96%
2-(Ethylcarbamoyl)pyridine-5-boronic acid pinacol ester

Purity: 96% CAS No.: 1006876-28-3 Molecular Formula: C14H21BN2O3 Formula Weight: 276.14 Storage Temperature: 2°C to 8°C Melting Point: 52-54° MDL No.: MFCD13191367

Formula: C₁₄H₂₁BN₂O₃
Melting Pt: 52…54 °C
Storage Temperature: Ambient
MDL Number: MFCD13191367
CAS Number: 1006876-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR