2-Ethylhexyl-4-(dimethylamino)benzoate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Ethylhexyl 4-(dimethylamino)benzoate

H54136-AE H54136-AP
AAH54136-AEEA 53.3 USD
AAH54136-AE AAH54136-AP
2-Ethylhexyl-4-(dimethylamino)benzoate 99%
2-Ethylhexyl 4-(dimethylamino)benzoate

Purity: 99% CAS No.: 21245-02-3 Molecular Formula: C17H27NO2 Formula Weight: 277.40 Flash Point: 113°(235°F) Refractive Index: 1.5420 Boiling Point: 325° Merck Reference: 14,3232 MDL No.: MFCD00017526

Formula: C₁₇H₂₇NO₂
MW: 277.40 g/mol
Boiling Pt: 325 °C
Density: 0.995
Flash Pt: 113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00017526
CAS Number: 21245-02-3
EINECS: 244-289-3
Merck Index: 14,3232

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR