3-Fluoro-4-(2-pyridylcarbamoyl)phenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Fluoro-4-(2-pyridylcarbamoyl)benzeneboronic acid
H54122-03
H54122-MD
AAH54122-03EA
424
USD
AAH54122-03
AAH54122-MD
3-Fluoro-4-(2-pyridylcarbamoyl)phenylboronic acid 97%
3-Fluoro-4-(2-pyridylcarbamoyl)phenylboronic acid
Purity: 97%
Molecular Formula: C12H10BFN2O3
Formula Weight: 260.03
MDL No.: MFCD20265181
Formula:
C₁₂H₁₀BFN₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD20265181 CAS Number: 1449133-22-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...