4-Cyanophenylacetonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H54112-03
H54112-06
AAH54112-03EA
53
USD
AAH54112-03
AAH54112-06
4-Cyanophenylacetonitrile 97%
4-Cyanophenylacetonitrile
Purity: 97%
CAS No.: 876-31-3
Molecular Formula: C9H6N2
Formula Weight: 142.16
Flash Point: 71°(160°F)
Refractive Index: 1.499
Boiling Point: 180-181°
MDL No.: MFCD03839848
Formula:
C₉H₆N₂ Boiling Pt: 180…181 °C Density: 1.056 Flash Pt: 71 °C(160 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00060305 CAS Number: 876-31-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...