[(tert-Butoxycarbonyl)aminooxy]acetic acid ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-Boc-(carboxymethoxy)amine, Boc-Aoa-OH, (Boc-aminooxy)acetic Acid
H54109-03
H54109-06
H54109-14
AAH54109-03EA
64.2
USD
AAH54109-03
AAH54109-06
AAH54109-14
[(tert-Butoxycarbonyl)aminooxy]acetic acid ≥98%
[(tert-Butoxycarbonyl)aminooxy]acetic acid
Purity: 98+%
CAS No.: 42989-85-5
Molecular Formula: C7H13NO5
Formula Weight: 191.18
Melting Point: ca 115° dec.
Beilstein Reference: 6137646
MDL No.: MFCD01632027
Formula:
C₇H₁₃NO₅ MW: 191.18 g/mol Melting Pt: ca 115 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD01632027 CAS Number: 42989-85-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...