6-Bromoquinolin-2(1H)-one 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H54095-03
H54095-06
AAH54095-03EA
56.9
USD
AAH54095-03
AAH54095-06
AAH54095-14
6-Bromoquinolin-2(1H)-one 96%
6-Bromoquinolin-2(1H)-one
Purity: 96%
CAS No.: 1810-66-8
Molecular Formula: C9H6BrNO
Formula Weight: 224.05
Storage Temperature: 2°C to 8°C
Melting Point:
Formula:
C₉H₆BrNO Melting Pt: >200 °C Storage Temperature: Ambient |
MDL Number:
MFCD08703169 CAS Number: 1810-66-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...