N,N'-Bis-Z-1-guanylpyrazole ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N,N'-Bis-Z-1-Guanylpyrazole
H54092-03
H54092-06
AAH54092-03EA
83.8
USD
AAH54092-03
AAH54092-06
N,N'-Bis-Z-1-guanylpyrazole ≥98%
N,N'-Bis-Z-1-guanylpyrazole
Purity: 98+%
CAS No.: 152120-55-3
Molecular Formula: C20H18N4O4
Formula Weight: 378.38
Melting Point: 87-91°
MDL No.: MFCD02683516
Formula:
C₂₀H₁₈N₄O₄ MW: 378.38 g/mol Melting Pt: 87…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD02683516 CAS Number: 152120-55-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...