2-(Trifluoromethyl)pyrimidine-5-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H54088-03
H54088-06
H54088-MD
AAH54088-03EA
164
USD
AAH54088-03
AAH54088-06
AAH54088-MD
2-(Trifluoromethyl)pyrimidine-5-carboxylic acid 97%
2-(Trifluoromethyl)pyrimidine-5-carboxylic acid
Purity: 97%
CAS No.: 306960-77-0
Molecular Formula: C6H3F3N2O2
Formula Weight: 192.10
Melting Point: 170-175°
MDL No.: MFCD09054851
Formula:
C₆H₃F₃N₂O₂ Melting Pt: 170…175 °C Storage Temperature: Ambient |
MDL Number:
MFCD09054851 CAS Number: 306960-77-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...