(1S,3R)-(-)-Camphoric acid 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (1S,3R)-1,2,2-Trimethylcyclopentane-1,3-dicarboxylic acid

H54042-03 H54042-06 H54042-MD
AAH54042-03EA 110 USD
AAH54042-03 AAH54042-06 AAH54042-MD
(1S,3R)-(-)-Camphoric acid 98%
(1S,3R)-(-)-Camphoric acid

Purity: 98% CAS No.: 560-09-8 Molecular Formula: C10H16O4 Formula Weight: 200.23 Melting Point: 188-190° Beilstein Reference: 1874674 Merck Reference: 14,1733 Optical Rotation: -48° (c=10 in ethanol) MDL No.: MFCD00064937

Formula: C₁₀H₁₆O₄
MW: 200.23 g/mol
Melting Pt: 188…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00064937
CAS Number: 560-09-8
Merck Index: 14,1733

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR