3-(4-tert-Butylphenyl)propionic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H54011-03 H54011-06 H54011-14
AAH54011-03EA 44.8 USD
AAH54011-03 AAH54011-06 AAH54011-14
3-(4-tert-Butylphenyl)propionic acid 97%
3-(4-tert-Butylphenyl)propionic acid

Purity: 97% CAS No.: 1208-64-6 Molecular Formula: C13H18O2 Formula Weight: 206.28 Melting Point: 102° MDL No.: MFCD00796482

Formula: C₁₃H₁₈O₂
Melting Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00796482
CAS Number: 1208-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR