5,7-Dichloro-1-indanone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5,7-Dichloro-2,3-dihydro-1H-inden-1-one
H54007-03
H54007-06
AAH54007-03EA
108
USD
AAH54007-03
AAH54007-06
5,7-Dichloro-1-indanone 97%
5,7-Dichloro-1-indanone
Purity: 97%
CAS No.: 448193-94-0
Molecular Formula: C9H6Cl2O
Formula Weight: 201.05
Melting Point: 98°
MDL No.: MFCD09908160
Formula:
C₉H₆Cl₂O MW: 201.05 g/mol Melting Pt: 98 °C Storage Temperature: Ambient |
MDL Number:
MFCD09908160 CAS Number: 448193-94-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...