5,7-Dibromo-1-indanone 97%

Supplier: Thermo Scientific Chemicals
Warning

H54005-03 H54005-06
AAH54005-03EA 96.6 USD
AAH54005-03 AAH54005-06
5,7-Dibromo-1-indanone 97%
5,7-Dibromo-1-indanone

Purity: 97% CAS No.: 923977-18-8 Molecular Formula: C9H6Br2O Formula Weight: 289.95 Melting Point: 148° MDL No.: MFCD11707100

Formula: C₉H₆Br₂O
MW: 289.95 g/mol
Melting Pt: 148 °C
Storage Temperature: Ambient
MDL Number: MFCD11707100
CAS Number: 923977-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR