3,3'-Diaminobenzidine (DAB) 99%

Supplier: Thermo Scientific Chemicals
Danger

H54000-06 H54000-14 H54000-22
AAH54000-06EA 69.75 USD
AAH54000-06 AAH54000-14 AAH54000-22
3,3'-Diaminobenzidine (DAB) 99%
3,3'-Diaminobenzidine (DAB)

Purity: 99% CAS No.: 91-95-2 Molecular Formula: C12H14N4 Formula Weight: 214.27 Storage Temperature: 2°C to 8°C Melting Point: 175-177° MDL No.: MFCD00007725

Formula: C₁₂H₁₄N₄
Melting Pt: 175…177 °C
Storage Temperature: Ambient
MDL Number: MFCD00007725
CAS Number: 91-95-2
EINECS: 202-110-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR