2-Bromo-4-(trifluoromethyl)phenylacetonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

H53518-03 H53518-06
AAH53518-03EA 56.8 USD
AAH53518-03 AAH53518-06 AAH53518-14
2-Bromo-4-(trifluoromethyl)phenylacetonitrile 98%
2-Bromo-4-(trifluoromethyl)phenylacetonitrile

Purity: 98% CAS No.: 474024-36-7 Molecular Formula: C9H5BrF3N Formula Weight: 264.04 Refractive Index: 1.5020 Boiling Point: 265-266° MDL No.: MFCD03094308

Formula: C₉H₅BrF₃N
MW: 264.04 g/mol
Boiling Pt: 265…266 °C
Density: 1.626
Storage Temperature: Ambient
MDL Number: MFCD03094308
CAS Number: 474024-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR