Ethyl-3-(trimethylsilyl)propiolate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 3-(trimethylsilyl)propynoate, 3-(Trimethylsilyl)-2-propynoic acid ethyl ester, 3-(Trimethylsilyl)propiolic acid ethyl ester, Ethyl 3-(trimethylsilyl)propiolate

H53517-06 H53517-14
AAH53517-06EA 61.2 USD
AAH53517-06 AAH53517-14
Ethyl-3-(trimethylsilyl)propiolate 98%
Ethyl-3-(trimethylsilyl)propiolate

Purity: 98% CAS No.: 16205-84-8 Molecular Formula: C8H14O2Si Formula Weight: 170.28 Flash Point: 71°(160°F) Refractive Index: 1.441 Boiling Point: 92-96°/28mm Beilstein Reference: 1767850 MDL No.: MFCD00074886

Formula: C₈H₁₄O₂Si
MW: 170.28 g/mol
Boiling Pt: 92…96 °C/28 mm
Density: 0.897
Flash Pt: 71 °C(160 °F)
Storage Temperature: Ambient
MDL Number: MFCD00074886
CAS Number: 16205-84-8
UN: 3334

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR