1-Chloro-5-(triethylsilyl)-4-pentyne 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H53515-06
H53515-14
AAH53515-06EA
48.8
USD
AAH53515-06
AAH53515-14
1-Chloro-5-(triethylsilyl)-4-pentyne 97%
1-Chloro-5-(triethylsilyl)-4-pentyne
Purity: 97%
CAS No.: 174125-30-5
Molecular Formula: C11H21ClSi
Formula Weight: 216.83
MDL No.: MFCD00671352
Formula:
C₁₁H₂₁ClSi Storage Temperature: Ambient |
MDL Number:
MFCD00671352 CAS Number: 174125-30-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...