3-Chloro-1-phenyl-1-propyne 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (3-Chloroprop-1-yn-1-yl)benzene

H53510-03 H53510-06 H53510-14
AAH53510-03EA 37.1 USD
AAH53510-03 AAH53510-06 AAH53510-14
3-Chloro-1-phenyl-1-propyne 97%
3-Chloro-1-phenyl-1-propyne

Purity: 97% CAS No.: 3355-31-5 Molecular Formula: C9H7Cl Formula Weight: 150.61 Flash Point: 104°(219°F) Refractive Index: 1.585 MDL No.: MFCD06411085

Formula: C₉H₇Cl
MW: 150.61 g/mol
Density: 1.095
Flash Pt: 104 °C(219 °F)
Storage Temperature: Ambient
MDL Number: MFCD06411085
CAS Number: 3355-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR