Rubrene 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
5,6,11,12-Tetraphenylnaphthacene, 5,6,11,12-Tetraphenyltetracene
H53507-03
H53507-06
H53507-14
AAH53507-03EA
84.3
USD
AAH53507-03
AAH53507-06
AAH53507-14
Rubrene 97%
Rubrene
Purity: 97%
CAS No.: 517-51-1
Molecular Formula: C42H28
Formula Weight: 532.68
Melting Point: 330-335°
Beilstein Reference: 1917339
Formula:
C₄₂H₂₈ MW: 532.68 g/mol Melting Pt: 330…335 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003703 CAS Number: 517-51-1 EINECS: 208-242-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...