cis-2-Hexene 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (Z)-2-Hexene

H53506-06 H53506-14
AAH53506-06EA 64.18 USD
AAH53506-06 AAH53506-14
cis-2-Hexene 96%
cis-2-Hexene

Purity: 96% CAS No.: 7688-21-3 Molecular Formula: C6H12 Formula Weight: 84.16 Flash Point: -25°(-13°F) Refractive Index: 1.396 Boiling Point: 68-70° Beilstein Reference: 1719019 UN No.: 3295 MDL No.: MFCD00066520

Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 68…70 °C
Melting Pt: -141.1 °C
Density: 0.6863 g/cm³ (20 °C)
Flash Pt: –25 °C (–13 °F)
Storage Temperature: Ambient
MDL Number: MFCD00066520
CAS Number: 7688-21-3
EINECS: 231-697-1
UN: 3295
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR