3-Bromobenzophenone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(3-Bromophenyl)(phenyl)methanone, m-Bromobenzophenone
H53505-03
H53505-06
H53505-14
AAH53505-03EA
69.8
USD
AAH53505-03
AAH53505-06
AAH53505-14
3-Bromobenzophenone 97%
3-Bromobenzophenone
Purity: 97%
CAS No.: 1016-77-9
Molecular Formula: C13H9BrO
Formula Weight: 261.12
Melting Point: 74-78°
MDL No.: MFCD00672015
Formula:
C₁₃H₉BrO MW: 261.12 g/mol Boiling Pt: 355 °C (1013 hPa) Melting Pt: 74…78 °C Density: 1.42 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00672015 CAS Number: 1016-77-9 EINECS: 213-808-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...