4-Diethylamino-2-butyn-1-ol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H53501-06
H53501-14
AAH53501-06EA
51.8
USD
AAH53501-06
AAH53501-14
4-Diethylamino-2-butyn-1-ol 98%
4-Diethylamino-2-butyn-1-ol
Purity: 98%
CAS No.: 10575-25-4
Molecular Formula: C8H15NO
Formula Weight: 141.21
Flash Point: 104°(219°F)
Refractive Index: 1.479
Boiling Point: 231-232°
MDL No.: MFCD00671356
Formula:
C₈H₁₅NO MW: 141.21 g/mol Boiling Pt: 231…232 °C Density: 0.952 Flash Pt: 104 °C(219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00671356 CAS Number: 10575-25-4 EINECS: 234-163-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...