3-Phenyl-1-propyne 97% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Propynylbenzene

H53500-06 H53500-14
AAH53500-06EA 123 USD
AAH53500-06 AAH53500-14
3-Phenyl-1-propyne 97% stabilized
3-Phenyl-1-propyne

Purity: 97% CAS No.: 10147-11-2 Molecular Formula: C9H8 Formula Weight: 116.16 Flash Point: 52°(126°F) Refractive Index: 1.526 Boiling Point: 75°/20mm UN No.: 3295 MDL No.: MFCD00134431

Formula: C₉H₈
MW: 116.16 g/mol
Boiling Pt: 75 °C/20 mm
Density: 0.934
Flash Pt: 52 °C (126 °F)
Storage Temperature: Ambient
MDL Number: MFCD00134431
CAS Number: 10147-11-2
UN: 3295
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR