3-Ethyl-3-oxetanemethanol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (3-Ethyloxetan-3-yl)methanol, 2-Hydroxymethyl-2-ethyl-1,3-epoxypropane, 3-Hydroxymethyl-3-ethyloxetane

H53497-06 H53497-14
AAH53497-06EA 37.9 USD
AAH53497-06 AAH53497-14
3-Ethyl-3-oxetanemethanol 97%
3-Ethyl-3-oxetanemethanol

3-Ethyl-3-oxetanemethanol is used as pharmaceutical intermediate.


Purity: 97% CAS No.: 3047-32-3 Molecular Formula: C6H12O2 Formula Weight: 116.16 Flash Point: 108°(226°F) Refractive Index: 1.453 Boiling Point: 96°/4mm MDL No.: MFCD00190143

Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 96 °C/4 mm
Density: 1.019
Flash Pt: 108 °C(226 °F)
Storage Temperature: Ambient
MDL Number: MFCD00190143
CAS Number: 3047-32-3
EINECS: 221-254-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR