(tert-Butyldimethylsilyl)acetylene 98%

Supplier: Thermo Scientific Chemicals
Danger

H53494-03 H53494-06 H53494-14
AAH53494-03EA 68.2 USD
AAH53494-03 AAH53494-06 AAH53494-14
(tert-Butyldimethylsilyl)acetylene 98%
(tert-Butyldimethylsilyl)acetylene

Purity: 98% CAS No.: 86318-61-8 Molecular Formula: C8H16Si Formula Weight: 140.30 Flash Point: 6°(43°F) Refractive Index: 1.451 Boiling Point: 130° Beilstein Reference: 1921466 UN No.: 1993 MDL No.: MFCD00191877

Formula: C₈H₁₆Si
MW: 140.3 g/mol
Boiling Pt: 130 °C
Density: 0.751
Flash Pt: 6 °C(43 °F)
Storage Temperature: Ambient
MDL Number: MFCD00191877
CAS Number: 86318-61-8
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR