N-(5-Hexynyl)phthalimide 97%

Supplier: Thermo Scientific Chemicals

H53489-06 H53489-14
AAH53489-06EA 76.5 USD
AAH53489-06 AAH53489-14
N-(5-Hexynyl)phthalimide 97%
N-(5-Hexynyl)phthalimide

Purity: 97% CAS No.: 6097-08-1 Molecular Formula: C14H13NO2 Formula Weight: 227.26 Melting Point: 68-72° MDL No.: MFCD00671372

Formula: C₁₄H₁₃NO₂
Melting Pt: 68…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00671372
CAS Number: 6097-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR