3,6-Di(2-pyridyl)-1,2,4,5-tetrazine 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H53484-03
H53484-06
H53484-14
AAH53484-03EA
49
USD
AAH53484-03
AAH53484-06
AAH53484-14
3,6-Di(2-pyridyl)-1,2,4,5-tetrazine 96%
3,6-Di(2-pyridyl)-1,2,4,5-tetrazine
Purity: 96%
CAS No.: 1671-87-0
Molecular Formula: C12H8N6
Formula Weight: 236.24
Melting Point: 225° dec.
MDL No.: MFCD00121717
Formula:
C₁₂H₈N₆ Melting Pt: 225 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00121717 CAS Number: 1671-87-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...