Benzyl propargyl ether 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
((Prop-2-yn-1-yloxy)methyl)benzene
H53481-06
H53481-14
AAH53481-06EA
41.2
USD
AAH53481-06
AAH53481-14
Benzyl propargyl ether 97%
Benzyl propargyl ether
Purity: 97%
CAS No.: 4039-82-1
Molecular Formula: C10H10O
Formula Weight: 146.19
Refractive Index: 1.5200
Boiling Point: 101-101°/18mm
MDL No.: MFCD00483987
Formula:
C₁₀H₁₀O Boiling Pt: 101…101 °C/18 mm Density: 1.011 Storage Temperature: Ambient |
MDL Number:
MFCD00483987 CAS Number: 4039-82-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...