2-(Trifluoromethyl)phenylacetylene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Ethynyl-2-(trifluoromethyl)benzene
H53478-03
H53478-06
AAH53478-03EA
56.9
USD
AAH53478-03
AAH53478-06
AAH53478-14
2-(Trifluoromethyl)phenylacetylene 97%
2-(Trifluoromethyl)phenylacetylene
Purity: 97%
CAS No.: 704-41-6
Molecular Formula: C9H5F3
Formula Weight: 170.13
Flash Point: 25°(77°F)
Refractive Index: 1.4760
Boiling Point: 45°/8mm
UN No.: 1993
MDL No.: MFCD00467313
Formula:
C₉H₅F₃ MW: 170.13 g/mol Boiling Pt: 45 °C/8 mm Density: 1.222 Flash Pt: 25 °C(77 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00467313 CAS Number: 704-41-6 UN: 1993 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...