6-Chloro-1-hexyne 98%

Supplier: Thermo Scientific Chemicals
Warning

H53476-AA H53476-AC
AAH53476-AAEA 56.8 USD
AAH53476-AA AAH53476-AC
6-Chloro-1-hexyne 98%
6-Chloro-1-hexyne

Purity: 98% CAS No.: 10297-06-0 Molecular Formula: C6H9Cl Formula Weight: 116.59 Flash Point: 37°(99°F) Refractive Index: 1.451 Boiling Point: 83°/95mm UN No.: 1993 MDL No.: MFCD00013697

Formula: C₆H₉Cl
Boiling Pt: 83 °C/95 mm
Density: 0.962
Flash Pt: 37 °C (99 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013697
CAS Number: 10297-06-0
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR