Propargyl ether 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dipropargylether

H53473-06 H53473-14
AAH53473-06EA 112.1 USD
AAH53473-06 AAH53473-14
Propargyl ether 98%
Propargyl ether

Purity: 98% CAS No.: 6921-27-3 Molecular Formula: C6H6O Formula Weight: 94.11 Flash Point: 24°(75°F) Refractive Index: 1.4430 Boiling Point: 55-57°/65mm UN No.: 3271 MDL No.: MFCD00048108

Formula: C₆H₆O
Boiling Pt: 55…57 °C/65 mm
Density: 0.914
Flash Pt: 24 °C(75 °F)
Storage Temperature: Ambient
MDL Number: MFCD00048108
CAS Number: 6921-27-3
UN: 3271
ADR: "3",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR