4-Phenyl-1-buten-4-ol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Phenyl-3-buten-1-ol

H53466-06 H53466-14
AAH53466-06EA 97.6 USD
AAH53466-06 AAH53466-14
4-Phenyl-1-buten-4-ol 97%
4-Phenyl-1-buten-4-ol

Purity: 97% CAS No.: 936-58-3 Molecular Formula: C10H12O Formula Weight: 148.20 Flash Point: 103°(217°F) Refractive Index: 1.53 Boiling Point: 234° MDL No.: MFCD00039617

Formula: C₁₀H₁₂O
MW: 148.20 g/mol
Boiling Pt: 234 °C
Density: 0.992
Flash Pt: 103 °C(217 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039617
CAS Number: 936-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR