3-Ethynylpyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

H53465-03 H53465-06 H53465-14
AAH53465-03EA 65.3 USD
AAH53465-03 AAH53465-06 AAH53465-14
3-Ethynylpyridine 97%
3-Ethynylpyridine

Purity: 97% CAS No.: 2510-23-8 Molecular Formula: C7H5N Formula Weight: 103.12 Flash Point: 57°(135°F) Melting Point: 39-40° Boiling Point: 83-84°/30mm UN No.: 1325 MDL No.: MFCD02177459

Formula: C₇H₅N
Boiling Pt: 83…84 °C/30 mm
Melting Pt: 39…40 °C
Flash Pt: 57 °C(135 °F)
Storage Temperature: Ambient
MDL Number: MFCD02177459
CAS Number: 2510-23-8
UN: 1325
ADR: "4.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR