6-Chloro-2-hexanone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Chlorohexan-2-one
H53462-06
H53462-14
AAH53462-06EA
62.3
USD
AAH53462-06
AAH53462-14
6-Chloro-2-hexanone 98%
6-Chloro-2-hexanone
Purity: 98%
CAS No.: 10226-30-9
Molecular Formula: C6H11ClO
Formula Weight: 134.61
Flash Point: 91°(196°F)
Refractive Index: 1.4435
Boiling Point: 86°/16mm
MDL No.: MFCD00191625
Formula:
C₆H₁₁ClO Boiling Pt: 86 °C/16 mm Density: 1.0348 g/cm³ (12 °C) Flash Pt: 91 °C(196 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00191625 CAS Number: 10226-30-9 EINECS: 233-546-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...