1,1,3-Triphenyl-2-propyn-1-ol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H53461-06
H53461-14
AAH53461-06EA
50.6
USD
AAH53461-06
AAH53461-14
1,1,3-Triphenyl-2-propyn-1-ol 97%
1,1,3-Triphenyl-2-propyn-1-ol
Purity: 97%
CAS No.: 1522-13-0
Molecular Formula: C21H16O
Formula Weight: 284.36
Melting Point: 80-82°
MDL No.: MFCD00004449
Formula:
C₂₁H₁₆O Melting Pt: 80…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004449 CAS Number: 1522-13-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...