Tripropargylamine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H53459-03
H53459-06
H53459-14
AAH53459-03EA
48.9
USD
AAH53459-03
AAH53459-06
AAH53459-14
Tripropargylamine 97%
Tripropargylamine
Purity: 97%
CAS No.: 6921-29-5
Molecular Formula: C9H9N
Formula Weight: 131.18
Flash Point: 57°(135°F)
Refractive Index: 1.484
Boiling Point: 79-85°/11mm
UN No.: 1993
MDL No.: MFCD00008577
Formula:
C₉H₉N Boiling Pt: 79…85 °C/11 mm Density: 0.927 Flash Pt: 57 °C(135 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008577 CAS Number: 6921-29-5 EINECS: 230-038-5 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...