5-Trimethylsilyl-4-pentyn-1-ol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-(Trimethylsilyl)pent-4-yn-1-ol

H53457-06 H53457-14
AAH53457-06EA 103 USD
AAH53457-06 AAH53457-14
5-Trimethylsilyl-4-pentyn-1-ol 97%
5-Trimethylsilyl-4-pentyn-1-ol

Purity: 97% CAS No.: 13224-84-5 Molecular Formula: C8H16OSi Formula Weight: 156.30 Flash Point: 84°(183°F) Refractive Index: 1.457 Boiling Point: 76°/15mm MDL No.: MFCD00190229

Formula: C₈H₁₆OSi
MW: 156.30 g/mol
Boiling Pt: 76 °C/15 mm
Density: 0.861
Flash Pt: 84 °C (183 °F)
Storage Temperature: Ambient
MDL Number: MFCD00190229
CAS Number: 13224-84-5
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR