3-Bromo-1-(trimethylsilyl)-1-propyne 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(Trimethylsilyl)propargyl bromide, (3-Bromoprop-1-yn-1-yl)trimethylsilane
H53451-06
H53451-14
AAH53451-06EA
131.48
USD
AAH53451-06
AAH53451-14
3-Bromo-1-(trimethylsilyl)-1-propyne 98%
3-Bromo-1-(trimethylsilyl)-1-propyne
Purity: 98%
CAS No.: 38002-45-8
Molecular Formula: C6H11BrSi
Formula Weight: 191.14
Flash Point: 63°(145°F)
Refractive Index: 1.483
Boiling Point: 44-45°/2mm
Beilstein Reference: 2037650
MDL No.: MFCD00134460
Formula:
C₆H₁₁BrSi Boiling Pt: 44…45 °C/2 mm Density: 1.17 Flash Pt: 63 °C(145 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00134460 CAS Number: 38002-45-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...