4-Phenyl-3-butyn-2-ol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H53450-06
H53450-14
AAH53450-06EA
92.2
USD
AAH53450-06
AAH53450-14
4-Phenyl-3-butyn-2-ol 97%
4-Phenyl-3-butyn-2-ol
Purity: 97%
CAS No.: 5876-76-6
Molecular Formula: C10H10O
Formula Weight: 146.19
Flash Point:
Formula:
C₁₀H₁₀ MW: 146.19 g/mol Density: 1.005 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00046651 CAS Number: 5876-76-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...