cis-5-Norbornene exo-2,3-dicarboxylic anhydride 95%

Supplier: Thermo Scientific Chemicals
Danger

H53446-03 H53446-06 H53446-14
AAH53446-03EA 55.6 USD
AAH53446-03 AAH53446-06 AAH53446-14
cis-5-Norbornene exo-2,3-dicarboxylic anhydride 95%
cis-5-Norbornene exo-2,3-dicarboxylic anhydride

Purity: 95% CAS No.: 2746-19-2 Molecular Formula: C9H8O3 Formula Weight: 164.16 Melting Point: 140-145° MDL No.: MFCD00630580

Formula: C₉H₈O₃
MW: 164.16 g/mol
Melting Pt: 140…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00630580
CAS Number: 2746-19-2
EINECS: 220-384-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR