5-Hexenyl acetate 97%

Supplier: Thermo Scientific Chemicals
Warning

H53445-06 H53445-14 H53445-22
AAH53445-06EA 35.2 USD
AAH53445-06 AAH53445-14 AAH53445-22
5-Hexenyl acetate 97%
5-Hexenyl acetate

Purity: 97% CAS No.: 5048-26-0 Molecular Formula: C8H14O2 Formula Weight: 142.20 Flash Point: 60°(142°F) Refractive Index: 1.423 Boiling Point: 173-174° UN No.: 3272 MDL No.: MFCD00060856

Formula: C₈H₁₄O₂
MW: 142.20 g/mol
Boiling Pt: 173…174 °C
Density: 0.883
Flash Pt: 60 °C (142 °F)
Storage Temperature: Ambient
MDL Number: MFCD00060856
CAS Number: 5048-26-0
UN: 3272
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR