Propargyl benzoate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Prop-2-yn-1-yl benzoate

H53444-06 H53444-14
AAH53444-06EA 46.8 USD
AAH53444-06 AAH53444-14
Propargyl benzoate 98%
Propargyl benzoate

Purity: 98% CAS No.: 6750-04-5 Molecular Formula: C10H8O2 Formula Weight: 160.17 Refractive Index: 1.5320 Boiling Point: 225-226° MDL No.: MFCD00996341

Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Boiling Pt: 225…226 °C
Density: 1.106
Storage Temperature: Ambient
MDL Number: MFCD00996341
CAS Number: 6750-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR