Allyloxytrimethylsilane 99%

Supplier: Thermo Scientific Chemicals
Danger

H53443-06 H53443-14
AAH53443-06EA 23.5 USD
AAH53443-06 AAH53443-14
Allyloxytrimethylsilane 99%
Allyloxytrimethylsilane

Purity: 99% CAS No.: 18146-00-4 Molecular Formula: C6H14OSi Formula Weight: 130.26 Flash Point: 1°(34°F) Refractive Index: 1.397 Boiling Point: 101-102° Beilstein Reference: 1739517 UN No.: 1993 MDL No.: MFCD00008643

Formula: C₆H₁₄OSi
MW: 130.26 g/mol
Boiling Pt: 101…102 °C
Density: 0.773
Flash Pt: 1 °C (34 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008643
CAS Number: 18146-00-4
EINECS: 242-031-4
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR