4-Methyl-2-pentyne 97%

Supplier: Thermo Scientific Chemicals
Danger

H53442-03 H53442-06 H53442-14
AAH53442-03EA 34.2 USD
AAH53442-03 AAH53442-06 AAH53442-14
4-Methyl-2-pentyne 97%
4-Methyl-2-pentyne

Purity: 97% CAS No.: 21020-27-9 Molecular Formula: C6H10 Formula Weight: 82.15 Flash Point: -12°(-10°F) Refractive Index: 1.407 Boiling Point: 71-73° UN No.: 3295 MDL No.: MFCD00041615

Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 71…73 °C
Density: 0.711
Flash Pt: -12 °C(-10 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041615
CAS Number: 21020-27-9
UN: 3295
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR