3-Methyl-1-butyne 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H53439-06
H53439-14
AAH53439-06EA
121
USD
AAH53439-06
AAH53439-14
3-Methyl-1-butyne 96%
3-Methyl-1-butyne
Purity: 97%
CAS No.: 598-23-2
Molecular Formula: C5H8
Formula Weight: 68.12
Storage Temperature: 2°C to 8°C
Flash Point: -30°(-22°F)
Refractive Index: 1.374
Boiling Point: 30°
UN No.: 3295
MDL No.: MFCD00039853
Formula:
C₅H₈ MW: 68.12 g/mol Boiling Pt: 30 °C Melting Pt: -90.1 °C Density: 0.667 g/cm³ (20 °C) Flash Pt: –30 °C (–22 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039853 CAS Number: 598-23-2 EINECS: 209-925-6 UN: 3295 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...