1-Phenyl-1-[4-(1-piperidinyl)phenyl]-2-propyn-1-ol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H53435-06
H53435-14
AAH53435-06EA
78.9
USD
AAH53435-06
AAH53435-14
1-Phenyl-1-[4-(1-piperidinyl)phenyl]-2-propyn-1-ol 97%
1-Phenyl-1-[4-(1-piperidinyl)phenyl]-2-propyn-1-ol
Purity: 97%
CAS No.: 214746-69-7
Molecular Formula: C20H21NO
Formula Weight: 291.39
Melting Point: 142-144°
MDL No.: MFCD17677347
Formula:
C₂₀H₂₁NO MW: 291.39 g/mol Melting Pt: 142…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD17677347 CAS Number: 214746-69-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...