N-(4-Pentynyl)phthalimide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Pent-4-yn-1-yl)isoindoline-1,3-dione

H53432-06 H53432-14
AAH53432-06EA 67.3 USD
AAH53432-06 AAH53432-14
N-(4-Pentynyl)phthalimide 97%
N-(4-Pentynyl)phthalimide

Purity: 97% CAS No.: 6097-07-0 Molecular Formula: C13H11NO2 Formula Weight: 213.24 Melting Point: 87-91° MDL No.: MFCD06798110

Formula: C₁₃H₁₁NO₂
Melting Pt: 87…91 °C
Storage Temperature: Ambient
MDL Number: MFCD06798110
CAS Number: 6097-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR