2-(2-Propynyl)-1H-isoindole-1,3(2H)-dione 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(Prop-2-ynyl)isoindoline-1,3-dione, N-Propargylphthalimide, N-(2-Propynyl)phthalimide
H53429-06
H53429-14
AAH53429-06EA
70.8
USD
AAH53429-06
AAH53429-14
2-(2-Propynyl)-1H-isoindole-1,3(2H)-dione 98%
2-(2-Propynyl)-1H-isoindole-1,3(2H)-dione
Purity: 98%
CAS No.: 7223-50-9
Molecular Formula: C11H7NO2
Formula Weight: 185.18
Melting Point: 148-152°
MDL No.: MFCD00065028
Formula:
C₁₁H₇NO₂ MW: 185.18 g/mol Melting Pt: 148…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00065028 CAS Number: 7223-50-9 EINECS: 230-621-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...